Learn More
GE Healthcare Dharmacon siSTABLE™ Non-Targeting siRNA
Brand:  GE Healthcare Dharmacon D-001700-01-05
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11577220
  • £150.22 / 5nmol
Estimated Shipment date
from Supplier 29-03-2018
Add to basket
Specifications
Specifications
Negative control for use in vivo and other experimental settings with high Nuclease activity, BLAST analysis confirms at least 4 mismatches with all known Human, Mouse and Rat genes
5nm
Videos