GE Healthcare Dharmacon siSTABLE™ Non-Targeting siRNA
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001700-01-20
SISTABLE NONTARG SIR 1 20NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11587220
Quantity Price
1 £ 413.66 / 20nmol
Estimated Shipment date
from Supplier 23-02-2017
Add to basket
Description and Specification
Specification
Quantity 20nm
For Use With (Application) Negative control for use in vivo and other experimental settings with high Nuclease activity, BLAST analysis confirms at least 4 mismatches with all known Human, Mouse and Rat genes
Quantity 20nm