Learn More
GE Healthcare Dharmacon™ Acell™ siRNA Reagents
For use with Acell siRNA.
Brand:  GE Healthcare Dharmacon™ B-003000-WB-100
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11557170
  • £19.98 / 100mL
Estimated Shipment date
from Supplier 30-01-2018
Add to basket
Description
Description
Ancillary reagents for experimental workflow include:
5X siRNA Buffer:
Appropriate for resuspension and long-term storage of any short, double-stranded or single-stranded synthetic RNA molecule, including modified and unmodified siRNAs, microRNA Mimics and microRNA Hairpin Inhibitors. Composition: 300mM KCl, 30mM HEPES-pH 7.5, 1.0mM MgCl2
RNase-free water:
Molecular Grade RNase-free water for dilution of 5x buffer
DharmaFECT Cell Culture Reagent:
DharmaFECT Cell Culture Reagent (DCCR) is used to dilute DharmaFECT siRNA transfection reagent prior to use in Dharmacon Reverse Transfection format (RTF) siRNA library plates
Specifications
Specifications
Dilution of 5x Buffer or resuspension of RNA, it is essential to maintain RNase-free conditions at all times when working with RNA as they are very susceptible to degradation from environmental RNases
RNase-free water
100mL
Videos