Learn More
GE Healthcare Dharmacon™ Acell™ siRNA Reagents
For use with Acell siRNA.
Brand:  GE Healthcare Dharmacon™ B-002000-UB-100
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 3.17510kg
 View more versions of this product
Product Code. 11547170
  • £61.42 / 100mL
In Stock 3
Add to basket
Specifications
Description
Ancillary reagents for experimental workflow include:
5X siRNA Buffer:
Appropriate for resuspension and long-term storage of any short, double-stranded or single-stranded synthetic RNA molecule, including modified and unmodified siRNAs, microRNA Mimics and microRNA Hairpin Inhibitors. Composition: 300mM KCl, 30mM HEPES-pH 7.5, 1.0mM MgCl2
RNase-free water:
Molecular Grade RNase-free water for dilution of 5x buffer
DharmaFECT Cell Culture Reagent:
DharmaFECT Cell Culture Reagent (DCCR) is used to dilute DharmaFECT siRNA transfection reagent prior to use in Dharmacon Reverse Transfection format (RTF) siRNA library plates
Description & Specifications
Specifications
Resuspension and long-term storage of any short, double-stranded or single-stranded synthetic RNA molecule, provides long-term storage of siRNAs, RNAs, microRNA reagents
5x siRNA buffer
100mL