GE Healthcare Dharmacon siGLO™ RISC-Free Control

Isolate cellular effects related only to siRNA transfection

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001600-01-20

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11527210

Quantity Price
1 £ 479.52 / 20nmol
Estimated Shipment date
from Supplier 04-04-2017
Add to basket


Description and Specification

Specification

Fluorophore Cy3 Rhodamine or Polyethylene
Product Type siGLO RISC-Free siRNA
Quantity 20nmol
Filter Type Cy3 Rhodamine or Polyethylene
For Use With (Application) Isolating cellular effects related only to siRNA transfection
Quantity 20nmol