Learn More
GE Healthcare Dharmacon siGLO™ Red Transfection Indicator
Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.
Brand:  GE Healthcare Dharmacon D-001630-02-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11567220
  • £392.20 / 20nmol
Estimated Shipment date
from Supplier 01-08-2017
Add to basket
Specifications Description & Specifications
Specifications
Cy3 Thodamine or Polyethylene
Determination of optimal siRNA transfection conditions in mammalian cells
20nm