Learn More
GE Healthcare Dharmacon siGLO™ Lamin A/C siRNA (Human/Mouse/Rat)
Brand:  GE Healthcare Dharmacon D-001620-01-20
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11567210
  • £479.52 / 20nmol
Estimated Shipment date
from Supplier 03-04-2018
Add to basket
Specifications
Specifications
RNAi experiments, positive correlation of cytoplasmic Fluorescence with transfection efficiency, silences the Cyclophilin B gene and is labeled with DY-547
20nm
Videos