GE Healthcare Dharmacon siGLO™ Lamin A/C siRNA (Human/Mouse/Rat)
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001620-01-05
SIGLO LAMINA/C SIR(HUMO) 5NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11557210
Quantity Price
1 £ 173.16 / 5nmol
Estimated Shipment date
from Supplier 27-02-2017
Add to basket
Description and Specification
Specification
Quantity 5nm
For Use With (Application) RNAi experiments, positive correlation of cytoplasmic Fluorescence with transfection efficiency, silences the Cyclophilin B gene and is labeled with DY-547
Quantity 5nm