Learn More
GE Healthcare Dharmacon siGLO™ Lamin A/C Control siRNA (Human)
Silences the human Lamin A/C gene and is labeled with DY-547
Brand:  GE Healthcare Dharmacon D-001620-03-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11597210
  • £173.16 / 5nmol
Estimated Shipment date
from Supplier 30-06-2017
Add to basket
Specifications Description & Specifications
Specifications
Cy3 Rhodamine or Polyethylene
NM_019390
5nmol
Lamin A/C siRNA - Mouse
RNAi experiments in Mouse cells, significantly extend siRNA stability and Fluorescence compared to conventional siRNAs similarly labeled with fluorophores