Learn More
GE Healthcare Dharmacon siGLO™ Lamin A/C Control siRNA (Human)
Silences the human Lamin A/C gene and is labeled with DY-547
Brand:  GE Healthcare Dharmacon D-001620-02-20
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11587210
  • £479.52 / 20nmol
Estimated Shipment date
from Supplier 26-02-2018
Add to basket
Specifications
Specifications
NM_005572, NM_170707, NM_170708
RNAi experiments in Human cells, significantly extend siRNA stability and Fluorescence compared to conventional siRNAs similarly labeled with fluorophores
20nmol
Cy3 Rhodamine or Polyethylene
Lamin A/C siRNA - Human
Videos