Learn More
GE Healthcare Dharmacon siGLO™ Lamin A/C Control siRNA (Human)
Silences the human Lamin A/C gene and is labeled with DY-547
Brand:  GE Healthcare Dharmacon D-001620-03-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11507220
  • £479.52 / 20nmol
Estimated Shipment date
from Supplier 01-09-2017
Add to basket
Specifications Description & Specifications
Specifications
Cy3 Rhodamine or Polyethylene
Lamin A/C siRNA - Mouse
RNAi experiments in Mouse cells, significantly extend siRNA stability and Fluorescence compared to conventional siRNAs similarly labeled with fluorophores
NM_019390
20nmol