GE Healthcare Dharmacon siGLO™ Green Transfection Indicator

Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001630-01-20

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11547220

Quantity Price
1 £ 392.2 / 20nmol
Estimated Shipment date
from Supplier 07-03-2017
Add to basket


Description and Specification

Specification

Fluorophore FITC
Quantity 20nm
Filter Type FITC
Quantity 20nm
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells