Learn More
GE Healthcare Dharmacon siGLO™ Green Transfection Indicator
Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.
Brand:  GE Healthcare Dharmacon D-001630-01-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11537220
  • £158.36 / 5nmol
Estimated Shipment date
from Supplier 06-06-2017
Add to basket
Specifications Description & Specifications
Specifications
FITC
Determination of optimal siRNA transfection conditions in mammalian cells
5nm