Learn More
GE Healthcare Dharmacon siGLO™ Cyclophilin B Control siRNA
Silences the Cyclophilin B gene and is labeled with DY-547
Brand:  GE Healthcare Dharmacon D-001610-01-20
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11547210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells
20nmol
Cy3 Thodamine or Polyethylene
siGLO Cyclophilin B siRNA
Videos