GE Healthcare Dharmacon siGLO™ Cyclophilin B Control siRNA

Silences the Cyclophilin B gene and is labeled with DY-547

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001610-01-05

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11537210

Quantity Price
1 £ 173.16 / 5nmol
Estimated Shipment date
from Supplier 30-03-2017
Add to basket


Description and Specification

Specification

Fluorophore Cy3 Thodamine or Polyethylene
Accession Number NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Product Type siGLO Cyclophilin B siRNA
Quantity 5nmol
For Use With (Application) Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells