Learn More
GE Healthcare Dharmacon siGLO™ Cyclophilin B Control siRNA
Silences the Cyclophilin B gene and is labeled with DY-547
Brand:  GE Healthcare Dharmacon D-001610-01-20
Code : P1
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 10066263
  • £479.52 / 20nmol
Estimated Shipment date
from Supplier 22-02-2018
Add to basket
Specifications
Specifications
NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells
20nmol
Cy3 Thodamine or Polyethylene
siGLO Cyclophilin B siRNA
Videos