GE Healthcare Dharmacon siGENOME™ RISC-Free Control
Effective RNAi transfection control that will not be taken up by RISC
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001220-01-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11557200
Quantity Price
1 £ 119.88 / 5nmol
Estimated Shipment date
from Supplier 04-05-2017
Add to basket
Description and Specification
Specification
Product Type RISC-Free siRNA #1
Quantity 5nmol
For Use With (Application) Isolating cellular effects related only to siRNA transfection