Learn More
GE Healthcare Dharmacon siGENOME™ RISC-Free Control
Effective RNAi transfection control that will not be taken up by RISC
Brand:  GE Healthcare Dharmacon D-001220-01-20
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11567200
  • £327.08 / 20nmol
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Specifications
Isolating cellular effects related only to siRNA transfection
RISC-Free siRNA #1
20nmol
Videos