Learn More
GE Healthcare Dharmacon siGENOME™ Non-Targeting siRNA Pools
Brand:  GE Healthcare Dharmacon D-001206-14-20
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11547190
  • £340.40 / 20nmol
Estimated Shipment date
from Supplier 02-03-2018
Add to basket
Specifications
Specifications
Determination of baseline cellular responses in RNAi experiments
20nm
Videos