GE Healthcare Dharmacon siGENOME™ Non-Targeting siRNA
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001210-04-20
SIGENOME NONTARG SIR 4 20NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11527200
Quantity Price
1 £ 319.68 / 20nmol
Estimated Shipment date
from Supplier 25-01-2017
Add to basket
Description and Specification
Specification
Quantity 20nm
For Use With (Application) Determination of baseline cellular responses in RNAi experiments, screened by microarray analysis
Quantity 20nm