Learn More
GE Healthcare Dharmacon siGENOME™ Non-Targeting Control siRNAs
Negative control siRNAs designed to target no known genes in human, mouse or rat
Brand:  GE Healthcare Dharmacon D-001210-01-20
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11567190
  • £319.68 / 20nmol
Estimated Shipment date
from Supplier 29-01-2018
Add to basket
Specifications
Specifications
Designed to have comparable GC content to that of functional siRNA, Used in siRNA-mediated RNAi experiments as controls for nonsequence specific effects and determination of baseline cellular responses in RNAi experiments
siCONTROL Nontargeting siRNA #1
20nmol
Videos