Learn More
GE Healthcare Dharmacon siGENOME™ Lamin A/C Control siRNA
Validated positive silencing control targeting the Lamin A/C genes in human, mouse and rat cell lines
Brand:  GE Healthcare Dharmacon D-001050-01-20
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11557180
  • £218.30 / 20nmol
Estimated Shipment date
from Supplier 20-02-2018
Add to basket
Specifications
Specifications
NM_005572, NM_170707, NM_170708 (Human), NM_019390 (Mouse) and NM_001002016 (Rat)
Positive silencing for validation of experimental design and detection methodologies in Human, Mouse and Rat cell lines
siCONTROL Lamin A/C siRNA (Human, Mouse, Rat)
20nmol
Videos