Learn More
GE Healthcare Dharmacon siGENOME™ Lamin A/C Control siRNA
Validated positive silencing control targeting the Lamin A/C genes in human, mouse and rat cell lines
Brand:  GE Healthcare Dharmacon D-001050-01-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11547180
  • £96.20 / 5nmol
Estimated Shipment date
from Supplier 30-06-2017
Add to basket
Specifications Description & Specifications
Specifications
siCONTROL RISC-Free siRNA #1
NM_005572, NM_170707, NM_170708 (Human), NM_019390 (Mouse) and NM_001002016 (Rat)
Positive silencing for validation of experimental design and detection methodologies in Human, Mouse and Rat cell lines
5nmol