GE Healthcare Dharmacon siGENOME™ GAPD Control siRNA
Validated positive silencing control targeting GAPD gene in human cell lines
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001140-01-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11597180
Quantity Price
1 £ 96.2 / 5nmol
Estimated Shipment date
from Supplier 07-06-2017
Add to basket
Description and Specification
Specification
Accession Number NM_002046
Product Type siGENOME GAPD Control siRNA, Human
Quantity 5nmol
For Use With (Application) Validated positive silencing control targeting the GAPD gene in Human cell lines, useful for determination of optimal RNAi experimental conditions