Learn More
GE Healthcare Dharmacon siGENOME™ GAPD Control siRNA
Validated positive silencing control targeting GAPD gene in human cell lines
Brand:  GE Healthcare Dharmacon D-001140-01-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11507190
  • £218.30 / 20nmol
Estimated Shipment date
from Supplier 29-08-2017
Add to basket
Specifications Description & Specifications
Specifications
NM_002046
siGENOME GAPD Control siRNA, Human
Validated positive silencing control targeting the GAPD gene in Human cell lines, useful for determination of optimal RNAi experimental conditions
20nmol