Learn More
GE Healthcare Dharmacon siGENOME™ Cyclophilin B Control siRNA
Validated positive silencing control targeting the Cyclophilin B (PPIB) gene in human, mouse/rat cell lines
Brand:  GE Healthcare Dharmacon D-001136-01-05
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11577180
  • £96.20 / 5nmol
Estimated Shipment date
from Supplier 22-02-2018
Add to basket
Specifications
Specifications
NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Positive silencing Cyclophilin B mRNA for validation of experimental design and detection methodologies in Human, useful in determination of optimal RNAi experimental conditions
siCONTROL Cyclophilin B siRNA
5nmol
Videos