GE Healthcare Dharmacon™ siCONTROL™ Complete Kits
Eleven cross-species, validated siRNA controls provided at 5nmol quantity
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: K-002800-C2-04
RNAI COMPLETE KIT W/RAT
UNSPSC: 12352200
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11602920
Quantity Price
1 £ 747.4 / Each
Estimated Shipment date
from Supplier 20-02-2017
Add to basket
Description and Specification
Specification
For Use With (Application) Provide positive silencing for validation of experimental design and detection methodologies
Quantity 5 nmol
Product Type Lamin A/C Rat
Quantity 5 nmol