GE Healthcare Dharmacon™ ON-TARGETplus™ Non-Targeting siRNAs
Ideal negative control for RNAi experiments in human, mouse, and rat cells.
Overview
Brand: GE Healthcare Dharmacon™
Manufacturer Part Number: D-001810-01-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.22680kg
Product Code. 11557230
Quantity Price
1 £ 141.34 / 5nmol
In Stock 3
Add to basket
Description and Specification
Specification
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
Product Type ON-TARGETplus Non-Targeting SiRNA#1
Quantity 5nmol
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Designed to leverage seed-region optimization
  • Chemically matched for use with ON-TARGETplus siRNA reagents