Learn More
GE Healthcare Dharmacon™ ON-TARGETplus™ Non-Targeting siRNAs
Ideal negative control for RNAi experiments in human, mouse, and rat cells.
Brand:  GE Healthcare Dharmacon™ D-001810-04-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11527240
  • £359.64 / 20nmol
Estimated Shipment date
from Supplier 28-06-2017
Add to basket
Specifications
Description
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Designed to leverage seed-region optimization
  • Chemically matched for use with ON-TARGETplus siRNA reagents
Description & Specifications
Specifications
Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
ON-TARGETplus Non-Targeting SiRNA#4
20nmol