GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting siRNAs
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001810-04-05
ONTARGETPLUS NONTARG SIR 4 5NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.22680kg
Product Code. 11517240
Quantity Price
1 £ 141.34 / 5nmol
Estimated Shipment date
from Supplier 27-02-2017
Add to basket
Description and Specification
Specification
Product Type ON-TARGETplus Non-Targeting SiRNA#4
Quantity 5nm
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
Quantity 5nm