GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting siRNAs
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001810-02-20
ONTARGETPLUS NONTARG SIR 2 20NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11587230
Quantity Price
1 £ 359.64 / 20nmol
Estimated Shipment date
from Supplier 31-03-2017
Add to basket
Description and Specification
Specification
Product Type ON-TARGETplus Non-Targeting SiRNA#2
Quantity 20nm
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
Quantity 20nm