Learn More
GE Healthcare Dharmacon™ ON-TARGETplus™ Non-Targeting siRNAs
Ideal negative control for RNAi experiments in human, mouse, and rat cells.
Brand:  GE Healthcare Dharmacon™ D-001810-01-20
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11567230
  • £359.64 / 20nmol
Estimated Shipment date
from Supplier 01-03-2018
Add to basket
Description
Description
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Designed to leverage seed-region optimization
  • Chemically matched for use with ON-TARGETplus siRNA reagents
Specifications
Specifications
Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
ON-TARGETplus Non-Targeting SiRNA#1
20nmol
Videos