Learn More
GE Healthcare Dharmacon™ ON-TARGETplus™ Non-Targeting Pool
Ideal negative control for RNAi experiments in human, mouse, and rat cells.
Brand:  GE Healthcare Dharmacon™ D-001810-10-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11547240
  • £394.42 / 20nmol
Estimated Shipment date
from Supplier 02-06-2017
Add to basket
Specifications
Description
  • ON-TARGETplus is designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • ON-TARGETplus modifications reduce potential off-targets
  • Pool of four non-targeting siRNAs
  • Recommended for determination of baseline cellular responses in RNAi experiments
Description & Specifications
Specifications
Determination of baseline cellular responses in RNAi experiments, screened by genome-wide microarray analysis for minimal off-targets
20nmol