Learn More
GE Healthcare Dharmacon™ ON-TARGETplus™ Non-Targeting Pool
Ideal negative control for RNAi experiments in human, mouse, and rat cells.
Brand:  GE Healthcare Dharmacon™ D-001810-10-05
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 10300934
  • £168.72 / 5nmol
Estimated Shipment
08-02-2018
Add to basket
Description
Description
  • ON-TARGETplus is designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • ON-TARGETplus modifications reduce potential off-targets
  • Pool of four non-targeting siRNAs
  • Recommended for determination of baseline cellular responses in RNAi experiments
Specifications
Specifications
Determination of baseline cellular responses in RNAi experiments, screened by genome-wide microarray analysis for minimal off-targets
5nmol
Videos