Learn More
GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control siRNA
Designed and modified to minimize off-target effects and improve silencing specificity
Brand:  GE Healthcare Dharmacon D-001830-01-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11587250
  • £255.30 / 20nmol
Estimated Shipment date
from Supplier 25-10-2017
Add to basket
Specifications
Specifications
Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions
20nm
Human