GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control siRNA
Designed and modified to minimize off-target effects and improve silencing specificity
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001830-02-20
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11507260
Quantity Price
1 £ 255.3 / 20nmol
Estimated Shipment date
from Supplier 06-06-2017
Add to basket
Description and Specification
Specification
Species Mouse
For Use With (Application) Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions
Quantity 20nm