GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control siRNA

Designed and modified to minimize off-target effects and improve silencing specificity

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001830-03-20

ONTARGETPLUS GAPD SIR-RAT 20NMOL

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11527260

Quantity Price
1 £ 255.3 / 20nmol
Estimated Shipment date
from Supplier 27-02-2017
Add to basket


Description and Specification

Specification

Species Rat
Host Species Rat
Quantity 20nm
Quantity 20nm
For Use With (Application) Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions