GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control Pool

Designed and modified to minimize off-target effects and improve silencing specificity

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001830-30-05

ONTARGETPLUS GAPD POOL-RAT 5NMOL

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11577260

Quantity Price
1 £ 168.72 / 5nmol
Estimated Shipment date
from Supplier 28-02-2017
Add to basket


Description and Specification

Specification

Species Rat
Host Species Rat
Quantity 5nm
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
Quantity 5nm