Learn More
GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control siRNA
Designed and modified to minimize off-target effects and improve silencing specificity
Brand:  GE Healthcare Dharmacon D-001820-01-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11567240
  • £255.30 / 20nmol
Estimated Shipment date
from Supplier 29-08-2017
Add to basket
Specifications Description & Specifications
Specifications
Silence Cyclophilin B Mrna, useful for determination of optimal RNAi conditions
20nm
Human