GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control siRNA

Designed and modified to minimize off-target effects and improve silencing specificity

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001820-02-05

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11577240

Quantity Price
1 £ 119.88 / 5nmol
Estimated Shipment date
from Supplier 05-05-2017
Add to basket


Description and Specification

Specification

Species Mouse
For Use With (Application) Silence Cyclophilin B Mrna, useful for determination of optimal RNAi conditions
Quantity 5nm