Learn More
GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
Brand:  GE Healthcare Dharmacon D-001820-20-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11537250
  • £168.72 / 5nmol
Estimated Shipment date
from Supplier 22-09-2017
Add to basket
Specifications
Specifications
Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
5nm
Mouse