GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001820-10-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11517250
Quantity Price
1 £ 168.72 / 5nmol
Estimated Shipment date
from Supplier 04-05-2017
Add to basket
Description and Specification
Specification
Species Human
Quantity 5nm
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells