ON-TARGETplus™ Cyclophilin B Control Pool

Designed and modified to minimize off-target effects and improve silencing specificity

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001820-30-20

ONTARGETPLUS CYCLO POOL-RAT 20NMOL

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11567250

Quantity Price
1 £ 394.42 / 20nmol
Estimated Shipment date
from Supplier 28-03-2017
Add to basket


Description and Specification

Specification

Quantity 20nm
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
Quantity 20nm
Species Rat
Host Species Rat
Includes Patented ON-TARGETplus modifications to minimize off-target effects
Storage Requirements –20°C