GE Healthcare Dharmacon miRIDIAN™ miRNA Mimic Transfection Control with Dy547
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: CP0045000105
MIRIDIAN MIMIC TRANSFECTION CTRL
UNSPSC: 41105324
Code: NEW
Additional Details:
Additional Details: Weight: 0.18080kg
Product Code. 11853334
Quantity Price
1 £ 175.38 / 5nmol
Estimated Shipment date
from Supplier 24-02-2017
Add to basket
Description and Specification
Specification
Quantity 5nm
For Use With (Application) No identifiable effects on miRNA function in multiple cell lines, transfection can be monitored and visualized by epifluorescence
Quantity 5nm