GE Healthcare Dharmacon miRIDIAN™ microRNA Hairpin Inhibitor Positive Controls


Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: IP0045000105

miRIDIAN microRNA Hairpin Inhibitor Transfection Control with Dy547, 5nmol

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.18080kg



Product Code. 11848034

Quantity Price
1 £ 166.5 / Each
Estimated Shipment date
from Supplier 24-02-2017
Add to basket


Description and Specification

Specification

Product Type Transfection Positive Control, miRNA
Target Human, mouse and rat cells
Quantity 5nm
For Use With (Application) miRIDIAN microRNA Hairpin Inhibitor Positive Control
Quantity 5nm