GE Healthcare Dharmacon miRIDIAN™ microRNA Hairpin Inhibitor Positive Controls


Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: IP0040000105

miRIDIAN microRNA Hairpin Inhibitor Positive Control 5nmol

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.18080kg



Product Code. 11818034

Quantity Price
1 £ 166.5 / Each
Estimated Shipment date
from Supplier 24-02-2017
Add to basket


Description and Specification

Specification

Product Type siRNA Positive Control
Target microRNA (miR-16)
Quantity 1 × 5nmol
For Use With (Application) miRIDIAN microRNA Hairpin Inhibitor Positive Control
Quantity 1 × 5nmol