Learn More
GE Healthcare Dharmacon miRIDIAN™ microRNA Hairpin Inhibitor Positive Controls
Brand:  GE Healthcare Dharmacon IP0040000120
UNSPSC : 12352200
Code : NEW
Additional Details : Weight : 0.18080kg
 View more versions of this product
Product Code. 11838034
  • £410.70 / Each
Estimated Shipment date
from Supplier 28-07-2017
Add to basket
Specifications Description & Specifications
Specifications
miRIDIAN microRNA Hairpin Inhibitor Positive Control
miRNA Positive Control
4 × 5nmol
microRNA (miR-16)