Learn More
GE Healthcare Dharmacon Mimic Housekeeping Positive Control
Brand:  GE Healthcare Dharmacon CP0030000120
UNSPSC : 41105324
Code : NEW
Additional Details : Weight : 0.18080kg
 View more versions of this product
Product Code. 11883324
  • £423.28 / 20nmol
Estimated Shipment date
from Supplier 26-10-2017
Add to basket
Specifications
Specifications
Delivery and function can be evaluated to determine transfection conditions in each cell line/type, monitor the function of a mimic molecule at the mRNA level using standard transcript quantification methods
20nm