GE Healthcare Dharmacon Mimic Housekeeping Positive Control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: CP0030000220
MIRIDIAN MIMIC + CTRL #2 RAT
UNSPSC: 41105324
Code: NEW
Additional Details:
Additional Details: Weight: 0.18080kg
Product Code. 11813334
Quantity Price
1 £ 423.28 / 20nmol
Estimated Shipment date
from Supplier 30-03-2017
Add to basket
Description and Specification
Specification
Quantity 20nm
For Use With (Application) Delivery and function can be evaluated to determine transfection conditions in each cell line/type, monitor the function of a mimic molecule at the mRNA level using standard transcript quantification methods
Quantity 20nm