GE Healthcare Dharmacon DharmaFECT™-1 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: T-2001-03

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 1.27460kg



Product Code. 11591731

Quantity Price
1 £ 351.5 / 1.50mL
In Stock 2
Add to basket


Description and Specification

Specification

Concentration 1mg/mL
Product Type DharmaFECT-1 Reagent
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
Quantity 1.5mL